missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD24 Monoclonal Antibody (SN3), FITC, Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA110153
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD24 is a two-chain glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed at multiple stages of B-cell development, beginning with the bone marrow pro-B-cell compartment and continuing through mature, surface Ig positive B-cells. Plasma cell expression is very low or negative. CD24 is also expressed on the majority of B-lineage acute lymphoblastic leukemias, B-cell CCLs and B-cell non-Hodgkin's lymphomas. CD24 may play a role in regulation of B-cell proliferation and maturation, and control of autoimmunity.Spécification
CD24 | |
Monoclonal | |
FITC | |
Cd24 | |
CD24; CD24 antigen; CD24 antigen (small cell lung carcinoma cluster 4 antigen); CD24 molecule; CD24A; CD24a antigen; cluster of differentiation 24; FLJ22950; FLJ43543; heat stable antigen; heat-stable antigen; HSA; Ly-52; Lymphocyte antigen 52; M1/69-J11D heat stable antigen; MGC75043; Nectadrin; nectadrin heat stable antigen; R13-Ag; Signal transducer CD24; Small cell lung carcinoma cluster 4 antigen; X62 heat stable antigen | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
100133941 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
SN3 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P25063 | |
Cd24 | |
Glycoproteins purified from human NALM-1 cell line. | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |