missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD244.1 Rat anti-Mouse, BUV661, Clone: C9.1, BD Biosciences™
Rat Monoclonal Antibody
Marque: BD Biosciences 750524
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD244.1 Monoclonal antibody specifically detects CD244.1 in Mouse samples. It is validated for Flow Cytometry
Spécification
| CD244.1 | |
| Monoclonal | |
| Brilliant Ultraviolet 661 | |
| 2B4 BALB Alloantigen; Cd244; Ly90; NAIL; Nmrk; SLAMF4 | |
| Mouse MHC-unrestricted killer hybridoma 7D4 | |
| RUO | |
| Mouse | |
| IgG2b κ |
| Flow Cytometry | |
| C9.1 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV661 under optimal conditions that minimize unconjugated dye and antibody. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu