Learn More
CD262 (DR5) Monoclonal Antibody (MD5-1), Functional Grade, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
Marque: Affymetrix eBioscience 16-5883-025
Description
The MD5-1 monoclonal antibody reacts with mouse DR5, also known as TRAIL-R2, Apo2, TRICK2, and KILLER. DR5 binds to TRAIL, activates NF-kappaB, and induces TRAIL-mediated apoptosis. DR5 mRNA is expressed broadly by normal tissue and human antigen is also expressed by some tumor cell lines. Applications Reported: The MD5-1 antibody has been reported for use in flow cytometric analysis. It has also been reported for use in functional assays. Applications Tested: The MD5-1 antibody has been tested by flow cytometric analysis of mouse DR5-transfected cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/ μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
CD262 (DR5) Monoclonal antibody specifically detects CD262 (DR5) in Mouse samples. It is validated for Flow Cytometry, Functional Assay
Spécification
| CD262 (DR5) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.2 | |
| Q9QZM4 | |
| Armenian Hamster | |
| 25 mg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry, Functional Assay | |
| MD5-1 | |
| Functional Grade | |
| TNFRSF10B | |
| apoptosis inducing protein TRICK2A/2B; apoptosis inducing receptor TRAIL-R2; CD262; cytotoxic TRAIL receptor-2; death domain containing receptor for TRAIL/Apo-2L; death receptor 5; DR5; dr-5; Fas-like protein; Killer; KILLER/DR5; KILLER/DR5 TRAIL death-inducing receptor; Ly98; MK; p53-regulated DNA damage-inducible cell death receptor(killer); sCD262; soluble CD262; TNF receptor superfamily member 10b; TNF-related apoptosis-inducing ligand receptor 2; TNFRSF10B; TRAIL receptor 2; TRAILR2; TRAIL-R2; TRICK2; TRICK2A; TRICK2B; TRICKB; tumor necrosis factor receptor superfamily member 10B; tumor necrosis factor receptor superfamily, member 10b; tumor necrosis factor receptor-like protein ZTNFR9; UNQ160/PRO186; ZTNFR9 | |
| Affinity chromatography | |
| RUO | |
| 21933 | |
| 4° C | |
| Liquid |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.