Learn More
Invitrogen™ CD271 (NGF Receptor) Monoclonal Antibody (ME20.4), Super Bright™ 436, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
411.00€
Spécification
| Antigène | CD271 (NGF Receptor) |
|---|---|
| Clone | ME20.4 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
The monoclonal antibody ME20.4 recognizes CD271, a 75 kDa, type I transmembrane protein that belongs to the low-affinity neurotrophin receptor and tumor necrosis factor receptor superfamily. CD271 is expressed by neurons, Schwann cells, melanocytes, mesen
Neurotrophic factors control the survival, differentiation and maintenance of neurons in the peripheral and central nervous systems, and of other neural crest-derived cell types. Developing sympathetic neurons are absolutely dependent upon nerve growth factor (NGF) during the period of target competition in vivo. Antibodies reacting specifically with NGFR p75 are useful tools in the detection and characterization of NGFR p75, to enhance our understanding of a wide range of phenomena in the development, plasticity and repair of the nervous system.Spécification
| CD271 (NGF Receptor) | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| IgG1 κ | |
| 4°C, store in dark, DO NOT FREEZE! |
| ME20.4 | |
| Flow Cytometry | |
| Super Bright 436 | |
| Mouse | |
| Human, Mouse, Rat, Canine, Rabbit, Monkey, Sheep, Pig | |
| Primary | |
| Antibody |