missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD279 (PD-1) Mouse anti-Human , Alexa Fluor 647, Clone: MIH4, BD Biosciences™
Mouse Monoclonal Antibody
Marque: BD Biosciences 566851
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Human samples. It is validated for Flow Cytometry
Spécification
| CD279 (PD-1) | |
| Monoclonal | |
| Alexa Fluor 647 | |
| PD1; hPD-1; hPD-l; PDCD1; PDC1; Programmed cell death 1; SLEB2 | |
| Human PD-1 Transfected Cell Line | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| MIH4 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor 647 under optimum conditions, and unreacted Alexa Fluor 647 was removed. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu