missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse anti-Human , PE, Clone: OKT3, BD Biosciences™
Mouse Monoclonal Antibody
Marque: BD Biosciences 566684
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD3 Monoclonal antibody specifically detects CD3 in Human samples. It is validated for Flow Cytometry
Spécification
| CD3 | |
| Monoclonal | |
| PE | |
| CD3E; CD3e; T-cell surface antigen T3/Leu-4 epsilon; T3E; TCRE | |
| Sheep Erythrocyte Rosette-purified Human T Cells | |
| RUO | |
| Human | |
| IgG2a κ |
| Flow Cytometry | |
| OKT3 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu