missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD300a/LMIR1 Antibody (172224), mFluor Violet 450 SE, Novus Biologicals™
Tous les produits Bio Techne ProduitsDescription
CD300a/LMIR1 Monoclonal antibody specifically detects CD300a/LMIR1 in Mouse samples. It is validated for Flow Cytometry, CyTOF
Spécification
Spécification
| Antigène | CD300a/LMIR1 |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Clone | 172224 |
| Conjugué | mFluor Violet 450 SE |
| Dilution | Flow Cytometry, CyTOF-ready |
| Formule | 50mM Sodium Borate |
| Alias de gène | CD300a antigenCMRF35H9, CD300a molecule, CLM-8, CMRF-35H, CMRF35-H, CMRF35H leukocyte immunoglobulin-like receptor, CMRF35-H9, CMRF-35-H9CD300 antigen-like family member A, CMRF35HIRp60, CMRF35-like molecule 8, IgSF12, IGSF12NK inhibitory receptor, Immunoglobulin superfamily member 12, Inhibitory receptor protein 60, IRC1, IRC1/IRC2, IRC2, Irp60, leukocyte membrane antigen |
| Espèces hôtes | Rat |
| Immunogène | Mouse myeloma cell line NS0-derived recombinant mouse CD300a/LMIR1, Leu28-Arg183, Accession # BAC80268 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?