missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD301 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA582781
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Immunogen sequence: CPVNWVEHQD SCYWFSHSGM SWAEAEKYCQ LKNAHLVVIN SREEQNFVQK YLGSAYTWMG LSDPEGAWKW VDGTDYATGF QNWKPGQPDD WQGHGLGGGE DCAHFHPDGR WND Highest antigen sequence indentity to the following orthologs: Mouse - 71%, Rat - 71%.
This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may function as a cell surface antigen. Two transcript variants encoding distinct isoforms have been identified for this gene.
Spécification
| CD301 | |
| Polyclonal | |
| Unconjugated | |
| CLEC10A | |
| CD301; CD301a; Clec10a; CLECSF13; CLECSF14; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 13 (macrophage-derived); C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 14 (macrophage-derived); C-type lectin domain family 10 member A; C-type lectin domain family 10, member A; C-type lectin superfamily member 14; HML; HML2; macrophage asialoglycoprotein-binding protein 1; macrophage galactose N-acetyl-galactosamine specific lectin 1; Macrophage galactose/N-acetylgalactosamine-specific lectin; Macrophage lectin 2; macrophage lectin 2 (calcium dependent); M-ASGP-BP-1; MGL; Mgl1; MMGL | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10462 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IUN9 | |
| CLEC10A | |
| Recombinant protein corresponding to Human CLEC10A. Recombinant protein control fragment (Product #RP-88573). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu