Learn More
CD32 Rat, PE-Cy7, Clone: 2.4G2, enQuireBio™
Rat Monoclonal Antibody
Marque:  EnquireBio QAB87-PE7-100ug
Code nomenclature Nacres: NA.46
Code produit. 15982588
  • 420.00€ / 100µg
Spécification
CD32
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
FCGR2b
Protein A/G Purified
RUO
14131
IgG2b
Flow Cytometry
2.4G2
PE-Cyanine7
XP_006496722
Rat
100 μg
Primary
Store at 2-4°C.