Learn More
Invitrogen™ CD321 (F11R) Monoclonal Antibody (WK9), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1586.00€ - 3585.00€
Spécification
| Antigène | CD321 (F11R) |
|---|---|
| Clone | WK9 |
| Concentration | 0.5 mg/mL |
| Classification | Monoclonal |
| Conjugué | Unconjugated |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|
17160732
|
Invitrogen™
14-9321-37 |
2 mg |
1586.00€
2mg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
|
17170732
|
Affymetrix eBioscience
14-9321-95 |
10 mg | |||||||
Description
CD321 (F11R, JAM-1, JAM-A) is a cell-surface adhesion molecule belonging to the Ig superfamily of receptors. CD321 is a 32-41 kD glycoprotein, possessing two extracellular Ig-like domains, a transmembrane region, and a short cytosolic tail. The protein assembles at the membrane as a homodimer, and can associate laterally with other membrane proteins including integrins. CD321 is expressed on a variety of cell types, including leukocytes, platelets, erythrocytes, epithelial and endothelial cells, and plays a role in mediating cell-cell contact. CD321 can be found in close proximity to several components of epithelial tight junctions, including ZO-1 and occludin. CD321 is also involved in reovirus entry into cells.
CD321 (F11R, JAM-1, JAM-A) is a cell-surface adhesion molecule belonging to the Ig superfamily of receptors. CD321 is a 32-41 kD glycoprotein, possessing two extracellular Ig-like domains, a transmembrane region, and a short cytosolic tail. The protein assembles at the membrane as a homodimer, and can associate laterally with other membrane proteins including integrins. CD321 is expressed on a variety of cell types, including leukocytes, platelets, erythrocytes, stem cells, epithelial and endothelial cells, and plays a role in mediating cell-cell contact. CD321 can be found in close proximity to several components of epithelial tight junctions, including ZO-1 and occludin. CD321 is involved in the maintenance of epithelial and endothelial barriers, hematopoiesis, angiogenesis, and tumor proliferation, invasion, and metastasis.Spécification
| CD321 (F11R) | |
| 0.5 mg/mL | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9Y624 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| WK9 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 50848 | |
| Primary | |
| 4°C | |
| F11r |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.