missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD34 Antibody (HPCA1/1806R), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Marque: Novus Biologicals NBP2-54355F
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD34 Monoclonal antibody specifically detects CD34 in Human, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
Spécification
| CD34 | |
| Monoclonal | |
| FITC | |
| CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule | |
| Recombinant full-length human CD34 protein (Uniprot: P28906) | |
| 100 μg | |
| Adaptive Immunity, Angiogenesis, B Cell Development and Differentiation Markers, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors | |
| 947 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| HPCA1/1806R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu