Learn More
CD365 (TIM1) Monoclonal Antibody (1D12-TIM1), PE, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
175.00€ - 421.00€
Spécification
| Antigène | CD365 (TIM1) |
|---|---|
| Clone | 1D12-TIM1 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|
16370604
|
Affymetrix eBioscience
12-3659-41 |
25 Tests | |||||||
16380604
![]() |
Invitrogen™
12-3659-42 |
100 Tests |
421.00€
100 pièces |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
Description: This 1D12-TIM1 monoclonal antibody recognizes human CD365 also known as Tim-1, Kim-1 or Havcr1. This antibody was generated using the immunoglobulin V domain of the extracellular part of the CD365 receptor, and is predicted to recognize all three haplotypes of human CD365. Clone 1D12-TIM1 has been shown to block entry of HCV virus into human hepatoma Huh7.5 cells in a dose dependent manner. It has been also shown to block the uptake of apoptotic cells by CD365-expressing 769-P cells. Applications Reported: This 1D12-TIM1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 1D12-TIM1 antibody has been pre-diluted and tested by flow cytometric analysis of A549 cells. This may be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser
CD365 also known as T-cell immunoglobulin and mucin domain 1 (Tim-1) is a type 1 surface glycoprotein that consists of extracellular N-terminal immunoglobulin V-like domain and a mucin domain, and a short cytoplasmic tail. The endogenous ligand for CD365 is Tim-4. In addition, CD365 is known to be a cellular receptor or co-receptor for a number of viruses including hepatitis A and C viruses, Ebola virus, Marburg virus, Dengue virus, Zika virus, West Nile virus, and Japanese encephalitis virus. The direct binding of the viruses by CD365 occurs via phosphatildyl-serine exposed on viral envelopes. Polymorphisms within human CD365 gene (HAVCR1) are associated with susceptibility to infection and the disease severity. CD365 has also been implicated in the T cell co-stimulation. CD365 is primarily expressed in epithelial cells. It is also highly expressed in A549, 769-P and Caco-2 and cell lines. CD365 exists also as a soluble protein generated by metalloproteinase mediated proteolytic cleavage of the surface protein.Spécification
| CD365 (TIM1) | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Q96D42 | |
| Havcr1 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| 1D12-TIM1 | |
| Flow Cytometry | |
| PE | |
| Mouse | |
| Human | |
| 26762 | |
| IgV domain of human CD365 | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Havcr1 |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.