Learn More
CD4 Antibody (CT-4), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP1-28267
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD4 Monoclonal antibody specifically detects CD4 in Avian, Chicken, Turkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Spécification
| CD4 | |
| Monoclonal | |
| FITC | |
| CD_antigen: CD4, CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
| Chicken thymocytes and Ig-negative blood leukocytes | |
| 0.25 mg | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers | |
| 920 | |
| Store at 4°C. Do not freeze. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| CT-4 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Avian, Chicken, Turkey | |
| Purified |