Learn More
CD4 Mouse, APC, Clone: OKT4, enQuireBio™
Mouse Monoclonal Antibody
Marque:  EnquireBio QAB11-APC-100Tests
Code nomenclature Nacres: NA.46
Code produit. 15981488
  • 222.60€ / 100 tests
Spécification
CD4
Monoclonal
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
CD4
Protein A/G Purified
RUO
920
IgG2b κ
Flow Cytometry
OKT4
APC
NP_001181945
Mouse
100 Tests
Primary
Store at 2-4°C.