Learn More
CD4 Rat, Biotin, Clone: GK1.5, enQuireBio™
Rat Monoclonal Antibody
Marque:  EnquireBio QAB8-B-100ug
Code nomenclature Nacres: NA.46
 Afficher plus de versions de ce produit
Code produit. 15982508
  • 169.05€ / 100µg
Spécification
CD4
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH7.2
Cd4
Protein A/G Purified
RUO
12504
IgG2b κ
Flow Cytometry
GK1.5
Biotin
NP_038516
Rat
100 μg
Primary
Store at 2-4°C.