Learn More
CD45 Mouse anti-Human, PE, Clone: HI30, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech PE-65109-100TESTS
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The CD45 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD45. This antibody recognizes human antigen. The CD45 antibody has been validated for the following applications: FC analysis. This antibody is pe.
Spécification
| CD45 | |
| Monoclonal | |
| 5 μL (0.125 μg)/Test | |
| Mouse | |
| Primary | |
| Human |
| Flow Cytometry | |
| HI30 | |
| PE | |
| 100 Tests | |
| 5788 | |
| IgG1 |