missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CD47 Antibody (magrolimab) - Humanized, Novus Biologicals™
Tous les produits Bio Techne ProduitsDescription
CD47 Monoclonal antibody specifically detects CD47 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Spécification
Spécification
| Antigène | CD47 |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Clone | magrolimab |
| Conjugué | Unconjugated |
| Formule | Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) |
| Numéro d’ordre du gène | Q08722 |
| Alias de gène | antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen |
| Espèces hôtes | Human |
| Immunogène | CD47 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?