Learn More
CD5 Rat, PE, Clone: 53-7.3, enQuireBio™
Rat Monoclonal Antibody
Marque:  EnquireBio QAB13-PE-100ug
Code nomenclature Nacres: NA.46
Code produit. 15971528
  • 288.75€ / 100µg
Spécification
CD5
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
Cd5
Protein A/G Purified
RUO
12507
IgG2a κ
Flow Cytometry
53-7.3
PE
XP_006526684
Rat
100 μg
Primary
Store at 2-4°C.