missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD57 Antibody (NK1/7565), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP3-20869AF700
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD57 Monoclonal antibody specifically detects CD57 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CD57 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| HNK1, HNK-1, Human Natural Killer-1, L2, LEU7, LEU-7 | |
| Recombinant fragment (around aa1-200) of human CD57 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer, Cell Biology, Immunology | |
| 27087 | |
| Store at 4°C in the dark. | |
| IgG2 λ |
| Immunohistochemistry (Paraffin) | |
| NK1/7565 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction