Learn More
CD58/LFA-3 Antibody (13G5), Novus Biologicals™
Rabbit Monoclonal Antibody
Marque: Novus Biologicals NBP3-26677-50ul
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD58/LFA-3 Monoclonal antibody specifically detects CD58/LFA-3 in Human samples. It is validated for Western Blot, ELISA
Spécification
| CD58/LFA-3 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 13G5 | |
| Western Blot 1:500-1:2000, ELISA | |
| Ag3, CD58 antigen, CD58 antigen, (lymphocyte function-associated antigen 3), CD58 molecule, FLJ23181, FLJ43722, LFA-3, LFA3ag3, lymphocyte function-associated antigen 3, Surface glycoprotein LFA-3 | |
| A synthesized peptide derived from Human CD58/LFA-3 [UniProt P19256] | |
| 50 μL | |
| Cell Biology, Cellular Markers, Immunology | |
| 965 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |