missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD79b Mouse anti-Human, Oligo, Clone: CB3-1, BD™ AbSeq ™
Mouse Monoclonal Antibody
Marque: BD Biosciences 940239
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD79b Monoclonal antibody specifically detects CD79b in Human samples. It is validated for Single Cell 3' Sequencing
Spécification
| CD79b | |
| Monoclonal | |
| Oligo | |
| Ig-beta; IGB; B29 | |
| Purified CD79αβ from Ramos B Cell Line | |
| RUO | |
| Human | |
| Purified |
| Single Cell 3' Sequencing | |
| Clone CB3-1 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. | |
| IgG1 κ |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu