missing translation for 'onlineSavingsMsg'
Learn More

CD80 Hamster anti-Mouse, PE, Clone: 16-10A1, BD

Hamster Monoclonal Antibody

Marque:  BD Biosciences 561955

Code nomenclature Nacres: NA.46

 Afficher plus de versions de ce produit

Code produit. 15801509

  • ONLINE EXCLUSIVE PRICE
    93.10€
    93.09€ / 1 pièce
    Économisez 0.01€
    0.01% de réduction

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Description

Description

The 16-10A1 antibody reacts with CD80 (B7-1). This member of the Ig superfamily, along with CD86 (B7-2), participates in T-cell co-stimulation via interactions with CD28 and CD152 (CTLA-4). CD80 has been reported to be constitutively expressed on dendritic cells, monocytes, and peritoneal macrophages; and it is inducible on B cells by various means, including activation by LPS, IL-4, and the cross-linking of surface Ig. Expression of CD80 has been reported to be greatly enhanced on splenic B cells following activation by LPS, with peak expression occurring between 48 and 72 hours. It has been reported that the activation of purified B cells with LPS can induce CD80 expression in as few as 18 hours. The 16-10A1 antibody has been reported to block binding of CTLA-4 Ig to CD80 and to block T-cell activation by Con A-elicited peritoneal exudate cells and CD80-transfected cell lines. However, 16-10A1 antibody alone is not able to block T-cell activation by antigen-presenting cells. CD86 (B7-2) is an alternate ligand for CD28 and CD152 (CTLA-4). Preliminary reports indicate that the 16-10A1 mAb may block the binding of rat anti-CD80 mAb clone 1G10 (Cat. No. 553368). In addition, it has been reported that the 16-10A1 antibody may cross-react with an activation antigen expressed on IFN-γ-activated alveolar macrophages of the dog.

TRUSTED_SUSTAINABILITY
Spécification

Spécification

CD80
Monoclonal
0.2mg/mL
Cd80; B7; B7-1; Cd28l; Ly-53; MIC17; TSA1
Hamster
Affinity Purified
RUO
Mouse
IgG2 κ
Flow Cytometry
16-10A1
PE
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse CD80 (B7) Transfected Cell Line
25 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis

For Research Use Only.