missing translation for 'onlineSavingsMsg'
Learn More

CD84/SLAMF5 Antibody (153-4D9), PE , Novus Biologicals™

Code produit 18632168 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
18632168 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18632168 Supplier Novus Biologicals Supplier No. NBP311578PE

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD84/SLAMF5 Monoclonal antibody specifically detects CD84/SLAMF5 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigène CD84/SLAMF5
Applications Flow Cytometry
Classification Monoclonal
Clone 153-4D9
Conjugué PE
Formule PBS
Alias de gène CD84 antigen, CD84 antigen (leukocyte antigen), CD84 molecule, Cell surface antigen MAX.3, DKFZp781E2378, hCD84, hly9-beta, leucocyte differentiation antigen CD84, leukocyte antigen CD84, Leukocyte differentiation antigen CD84, mCD84, Signaling lymphocytic activation molecule 5, SLAM family member 5, SLAMF5LY9B
Espèces hôtes Mouse
Immunogène MLR-stimulated lymphocytes
Méthode de purification Protein A or G purified
Quantité 0.1 mL
État réglementaire RUO
Disciplines de recherche CD Markers, Immunology, Stem Cells
Primaire ou secondaire Primary
Identification génétique (Entrez) 8832
Espèces cibles Human
Contenu et stockage Store at 4°C in the dark.
Forme Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.