Learn More
CD86 Mouse anti-Human, FITC, Clone: K86, Abnova™
Mouse monoclonal antibody raised against human CD86.
Marque: Abnova MAB12416.50Reactions
Description
This gene encodes a type I membrane protein that is a member of the immunoglobulin superfamily. This protein is expressed by antigen-presenting cells, and it is the ligand for two proteins at the cell surface of T cells, CD28 antigen and cytotoxic T-lymphocyte-associated protein 4. Binding of this protein with CD28 antigen is a costimulatory signal for activation of the T-cell. Binding of this protein with cytotoxic T-lymphocyte-associated protein 4 negatively regulates T-cell activation and diminishes the immune response. Alternative splicing results in two transcript variants encoding different isoforms. Additional transcript variants have been described, but their full-length sequences have not been determined. [provided by RefSeq]
Spécification
| CD86 | |
| Monoclonal | |
| FITC | |
| Flow cytometry Immunofluorescence The optimal working dilution should be determined by the end user. | |
| B7-2/B70/CD28LG2/LAB72/MGC34413 | |
| Mouse | |
| Protein A/G affinity chromatography | |
| RUO | |
| 942 | |
| Store at 4°C. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Immunofluorescence | |
| K86 | |
| Mouse monoclonal antibody raised against human CD86. | |
| In PBS, pH 7.4 (0.09% sodium azide, 0.2% BSA) | |
| CD86 | |
| CD86 | |
| 50 Reactions | |
| Primary | |
| Human | |
| Liquid |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.