Learn More
Invitrogen™ CD88 (C5aR1) Monoclonal Antibody (S5/1), PE, eBioscience™

Mouse Monoclonal Antibody
Marque: Invitrogen™ 12-0888-42
Description
Description: This S5/1 monoclonal antibody recognizes human CD88 also known as C5aR1. C5aR1 is a complement receptor widely expressed on cells of the myeloid lineage. Clone S5/1 has been reported to block C5a binding. Applications Reported: This S5/1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This S5/1 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.
CD88, also known as C5aR1, is a C5a Anaphylatoxin Receptor that mediates the effects of one of the most potent chemotactic agents known. C5AR1 has been implicated in asthma, and its expression has been reported in myeloid blood cells, brain, liver, lung, spleen, heart, kidney, and intestinal tract. CD88 is the receptor for the chemotactic and inflammatory peptide anaphylatoxin C5a. Functionally, CD88 stimulates chemotaxis, granule enzyme release and superoxide anion production. CD88 is also a component of the classical pathway of the complement cascade; it may be involved with tissue injury associated with ischemia or sepsis and it may be a component of the membrane attack complex. Diseases associated with CD88 include Mannose-Binding Lectin Deficiency and Systemic Lupus Erythematosus.
Spécification
| CD88 (C5aR1) | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| P21730 | |
| C5AR1 | |
| BSA-Conjugated Peptide aa. 1-31. | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| Flow Cytometry | |
| S5/1 | |
| PE | |
| C5AR1 | |
| anaphylatoxin C5a receptor; C5A; C5a anaphylatoxin chemotactic receptor; C5a anaphylatoxin chemotactic receptor 1; C5a anaphylatoxin receptor; C5a ligand; C5a Receptor; C5AR; C5a-R; C5AR1; C5r1; Cd88; CD88 antigen; complement C5a receptor; complement C5a receptor 1; complement component 5 receptor 1; complement component 5, receptor 1; complement component 5a receptor 1; D7Msu1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 728 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.