Learn More
Novus Biologicals™ CD9 Antibody (Genentech patent anti-CD9) - Humanized, Novus Biologicals™
Code produit 30048086 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
1 mg
100 μg
50 μg
Conditionnement:
100µg
1mg
50µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30048086 1 mg 1mg
30047879 100 μg 100µg
30044997 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 30048086 Fournisseur Novus Biologicals™ Code fournisseur NBP3280881mg
Human Monoclonal Antibody
CD9 Monoclonal antibody specifically detects CD9 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Spécification
Antigène CD9
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone Genentech patent anti-CD9
Conjugué Unconjugated
Formule Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
Numéro d’ordre du gène P21926
Alias de gène BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24)
Espèces hôtes Human
Immunogène CD9
Méthode de purification Protein A purified
Quantité 1 mg
État réglementaire RUO
Disciplines de recherche Cancer, Stem Cell Markers
Primaire ou secondaire Primary
Identification génétique (Entrez) 928
Reconstitution Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Espèces cibles Human
Contenu et stockage Store at -20°C in powder form. Store at -80°C once reconstituted.
Forme Purified
Isotype IgG1