Learn More
CD90 (Thy-1) Mouse anti-Human, Brilliant Violet 605, Clone: 5E10, BD OptiBuild™
Mouse Monoclonal Antibody
Marque: BD Biosciences 747750
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD90 (Thy-1) Monoclonal antibody specifically detects CD90 (Thy-1) in Human samples. It is validated for Flow Cytometry.
Spécification
| CD90 (Thy-1) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| CD90 (Thy-1) | |
| Refrigerate (2 to 8°C) |
| Flow Cytometry | |
| 5E10 | |
| Brilliant Violet 605 | |
| 50 μg | |
| Primary | |
| Human | |
| IgG1 κ |