missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD98 Chimeric Recombinant Rabbit Monoclonal Antibody (EP3-1)

Rabbit Recombinant Monoclonal Antibody
Marque: Invitrogen™ MA551735
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Shown to bind to epitopes of sequences; LLHGDFHAFSAG-GGGC-NH2, LLHGDFH-GGGC-HN2, and FHAFSAG-GGGC-NH2, but showed no reactivity to live HeLa cells by IF.
CD98 (4F2) is a type II transmembrane glycoprotein which serves as the heavy chain of the heterodimeric amino acid transporters (HATs). CD98, linked to various light chains by disulfide bond, is responsible for cell surface expression and basolateral localization of this transporter complex in polarized epithelial cells and also interacts with beta1 integrins and increases their affinity for ligand. Besides its roles in amino acid transport, CD98 is thus involved in cell fusion and activation. It is implicated in regulation of cellular differentiation, growth and apoptosis.
Spécification
| CD98 Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| P08195 | |
| SLC3A2 | |
| Immunised mice and constructed IgG1 Fab phage display libraries from the spleen cells. CD98-reactive recombinant Fab fragments were retrieved by epitope peptide panning. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG κ |
| Flow Cytometry, Immunoprecipitation, T-Cell Activation, Immunocytochemistry | |
| EP3-1 | |
| Unconjugated | |
| SLC3A2 | |
| 4F2; 4F2 cell-surface antigen heavy chain; 4F2 heavy chain antigen; 4F2hc; 4T2HC; AI314110; antigen defined by monoclonal antibody 4F2, heavy chain; antigen identified by monoclonal antibodies 4F2; antigen identified by monoclonal antibodies 4F2, TRA1.10, TROP4, and T43; Cd98; CD98 antigen; CD98 heavy chain; CD98HC; Ly10; Ly-10; Ly-m10; Lymphocyte activation antigen 4F2 large subunit; MDU1; Mgp-2hc; monoclonal antibody 44D7; NACAE; RGD:3073}; SLC3A2; slc3a2 {ECO:0000312; solute carrier family 3 (activators of dibasic and neutral amino acid transport), member 2; solute carrier family 3 (amino acid transporter heavy chain), member 2; solute carrier family 3 member 2; solute carrier family 3, member 2; type II transmembrane protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 6520 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu