Learn More
CEACAM8/CD66b Antibody (G10F5), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
Marque:  Novus Biologicals NB100-77808
Code nomenclature Nacres: NA.46
Code produit. 18787583
  • ONLINE EXCLUSIVE PRICE
    409.00€
    386.40€ / 0.10mg
    Économisez 22.60€
    5% de réduction
Description
CEACAM8/CD66b Monoclonal specifically detects CEACAM8/CD66b in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunocytochemistry, CyTOF-ready.
For Research Use Only