missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Centaurin alpha-1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA552900
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Immunogen sequence: LGVFICLSCS GIHRNIPQVS KVKSVRLDAW EEAQVEFMAS HGNDAARARF ESKVPSFYYR PTPSDCQLLR EQWIRAKYER QEFIYPEKQE PYSAGYREGF LWKRG Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 89%.
Centaurin alpha-1 (ADAP1, ArfGAP With Dual PH Domains 1) is a Protein Coding gene. Among its related pathways are B Cell Receptor Signaling Pathway (sino) and Arf6 signaling events. Gene Ontology (GO) annotations related to this gene include GTPase activator activity and inositol 1,3,4,5 tetrakisphosphate binding. An important paralog of this gene is ADAP2.
Spécification
| Centaurin alpha-1 | |
| Polyclonal | |
| Unconjugated | |
| ADAP1 | |
| 4930431P11Rik; ADAP1; Arf-GAP with dual PH domain-containing protein 1; ArfGAP with dual PH domains 1; CENTA1; centaurin, alpha 1; centaurin-alpha; centaurin-alpha-1; Cnt-a1; GC1L; GCS1L; inositol(1,3,4,5)tetrakisphosphate receptor; p42IP4; phosphatidylinositol-3,4,5-triphosphate binding protein; putative MAPK-activating protein PM25 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11033 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75689 | |
| ADAP1 | |
| Recombinant protein corresponding to Human Centaurin alpha-1. Recombinant protein control fragment (Product #RP-89650). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu