Learn More
ChemR23/CMKLR1 Antibody (84939), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals FAB362PECY55
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
ChemR23/CMKLR1 Monoclonal antibody specifically detects ChemR23/CMKLR1 in Human samples. It is validated for Flow Cytometry
Spécification
| ChemR23/CMKLR1 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 84939 | |
| Flow Cytometry | |
| CHEMERINR, chemokine receptor-like 1, chemokine-like receptor 1, CHEMR23, DEZ, G-protein coupled receptor ChemR23, G-protein coupled receptor DEZ, MGC126105, MGC126106, orphan G-protein coupled receptor, Dez | |
| NS0 mouse myeloma cell line transfected with human ChemR23, Met1-Leu371, Accession # NP_004063 | |
| 0.5 mL | |
| GPCR, Immunology | |
| 1240 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG3 |