Learn More
CHGA, anti-Human, Clone: CHGA/1815R, Abnova™
Monoclonal Antibody
Marque: Abnova RAB00323.100ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
| CHGA | |
| Monoclonal | |
| Unconjugated | |
| In 10 mM PBS (0.05% BSA, 0.05% sodium azide). | |
| CGA | |
| Original antibody is raised against recombinant protein corresponding to full length human CHGA. | |
| RUO | |
| 1113 | |
| Store at 4°C. | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| CHGA/1815R | |
| Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| chromogranin A (parathyroid secretory protein 1) | |
| CHGA | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG κ |