missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA571396
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CHL1 Polyclonal Antibody for Western Blot
The protein encoded by this gene is a member of the L1 gene family of neural cell adhesion molecules. It is a neural recognition molecule that may be involved in signal transduction pathways. The deletion of one copy of this gene may be responsible for mental defects in patients with 3p- syndrome. Several alternatively spliced transcript variants of this gene have been described, but their full length nature is not known.
Spécification
| CHL1 | |
| Polyclonal | |
| Unconjugated | |
| CHL1 | |
| A530023M13Rik; AI465420; CALL; cell adhesion molecule L1 like; cell adhesion molecule L1-like; cell adhesion molecule with homology to L1CAM; cell adhesion molecule with homology to L1CAM (close homolog of L1); cell adhesion molecule with homology to L1CAM (close homologue of L1); Chl1; Chl1-like protein; close homolog of L1; L1 cell adhesion molecule 2; L1CAM2; LICAM2; neural cell adhesion molecule L1-like protein; Processed neural cell adhesion molecule L1-like protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10752 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O00533 | |
| CHL1 | |
| synthetic peptide directed towards the N terminal region of human CHL1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu