Learn More
CHL1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25250-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The protein encoded by this gene is a member of the L1 gene family of neural cell adhesion molecules. It is a neural recognition molecule that may be involved in signal transduction pathways. The deletion of one copy of this gene may be responsible for mental defects in patients with 3p- syndrome. Several alternatively spliced transcript variants of this gene have been described, but their full length nature is not known.Spécification
| CHL1 | |
| Polyclonal | |
| Unconjugated | |
| CHL1 | |
| CALL, CHL1, Close homolog of L1, L1CAM2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10752, 12661 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O00533, P70232 | |
| CHL1 | |
| CHL1 Fusion Protein Ag19263 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |