missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Chlamydia trachomatis MOMP Monoclonal Antibody (CL12-666.33.3), Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen™ MA5-46112
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
MA1-10660 detects Chlamydia trachomatis MOMP from C. trachomatis serovars A, B, Ba, C, D, E, F, G, H, I, J, K, L1, L2, and L3 samples. MA1-10660 has been successfully used in ELISA applications. The MA1-10660 immunogen is C. trachomatis elementary bodies.
Chlamydia trachomatis is an intracellular organism. Chlamydia usually infects the cervix and fallopian tubes of women and the urethra of men. Chlamydial infections are believed to be one of the most common of all STDs. It is generally thought that in a population of 15 million, there are up to 300,000 cases of chlamydia each year. Thus, there are many undiagnosed cases of chlamydia in the community. It has been estimated that the true prevalence of chlamydia in the sexually active population may be in the order of 5% to 10%. Chlamydia is one of the leading causes of blindness in underdeveloped countries.
Spécification
| Chlamydia trachomatis MOMP | |
| Monoclonal | |
| 0.4 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Mouse | |
| PEG precipitation | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG3 |
| ELISA | |
| CL12-666.33.3 | |
| Unconjugated | |
| C. trachomatis MOMP; Chlamydia MOMP | |
| C. trachomatis elementary bodies. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu