missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Chlorpyrifos Monoclonal Antibody (3E4)

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA549719
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Antibody tested in indirect ELISA.
Chlorpyrifos is an insecticide belonging to the group of organophosphothioates. It is used in the culture of vegetables and in citrus orchards. Chlorpyrifos is an acetylcholinesterase inhibitor and as such having a highly toxic effect on the nervous system. There is evidence that chlorpyrifos has an effect on the foetus, which may lead to behavioural disorders.
Spécification
| Chlorpyrifos | |
| Monoclonal | |
| 2.9 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Conjugate of CPF hapten and KLH. | |
| 200 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry, Lateral Flow, SDS-Page | |
| 3E4 | |
| Unconjugated | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering