Learn More
Chromogranin A / CHGA (Neuroendocrine Marker) Mouse, Clone: rCHGA/798, enQuireBio™
Mouse Monoclonal Antibody
Marque: EnquireBio 1113-MSM17-P0
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
| Chromogranin A / CHGA (Neuroendocrine Marker) | |
| Finely granular cytoplasmic | |
| rCHGA/798 | |
| Unconjugated | |
| Mouse | |
| 68-75kDa | |
| Protein A/G purified from Bioreactor concentrate. | |
| RUO | |
| 1113 | |
| IgG1 κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant full-length human CHGA protein | |
| PC12 cells. Adrenal gland, bowel, thyroid, pancreas, or pheochromocytoma. | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |