Learn More
PeproTech CLEC11A Polyclonal Antibody, PeproTech®, Invitrogen™

Goat Polyclonal Antibody
Marque: PeproTech 500-P99G-1MG
Description
AA Sequence of recombinant protein: MGHGARGAER EWEGGWGGAQ EEEREREALM LKHLQEALGL PAGRGDENPA GTVEGKEDWE MEEDQGEEEE EEATPTPSSG PSPSPTPEDI VTYILGRLAG LDAGLHQLHV RLHALDTRVV ELTQGLRQLR NAAGDTRDAV QALQEAQGRA EREHGRLEGC LKGLRLGHKC FLLSRDFEAQ PSASPHPLSP DQPNGGTLEN CVAQASDDGS WWDHDCQRRL YYVCEFPF. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human SCGF-beta. Anti-Human SCGF-beta-specific antibody was purified by affinity chromatography employing an immobilized Human SCGF-beta matrix. Sandwich ELISA: To detect Human SCGF-beta by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human SCGF-beta (500-P99GBt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human SCGF-beta. Western Blot: To detect Human SCGF-beta by Western Blot analysis this antibody can be used at a concentration of 0.1- 0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Human SCGF-beta is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
This gene encodes a member of the C-type lectin superfamily. The encoded protein is a secreted sulfated glycoprotein and functions as a growth factor for primitive hematopoietic progenitor cells. An alternative splice variant has been described but its biological nature has not been determined.
Spécification
| CLEC11A | |
| Polyclonal | |
| Unconjugated | |
| CLEC11A | |
| AW457320; b SCGF; beta SCGF; betaSCGF; bSCGF; Clec11a; CLECSF3; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 3; C-type lectin domain family 11 member A; C-type lectin domain family 11, member A; C-type lectin superfamily member 3; hCG_22930; LSLCL; lymphocyte secreted C-type lectin; lymphocyte secreted long form of C-type lectin; Osteolectin; P47; SCGF; SCGF b; SCGF beta; SCGF β; SCGFb; SCGFbeta; SCGFβ; stem cell growth factor; stem cell growth factor; lymphocyte secreted C-type lectin; β SCGF; βSCGF | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 6320 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| Q9Y240 | |
| CLEC11A | |
| E.coli-derived Recombinant Human SCGF-beta. | |
| 1 mg | |
| Primary | |
| Human | |
| Antibody |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.