Learn More
Invitrogen™ CLEC4E Monoclonal Antibody (WEN43)

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA533341
Description
Product is supplied at 1 mL (100 μL/mL). Application notes: Flow Cytometry - This anitbody stains the extracellular domain of Mincle transfected 293 cells. It does not bind to other receptors in the rat APLEC family. Functional Assay - This antibody can act complementary in the inhibition of phagocytosis. Positive control: BW5417 or 293T cells.
CLEC4E encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signaling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type II transmembrane protein is a downstream target of CCAAT/enhancer binding protein (C/EBP), beta (CEBPB) and may play a role in inflammation. Alternative splice variants have been described but their full-length sequence has not been determined. This gene is closely linked to other CTL/CTLD superfamily members on chromosome 12p13 in the natural killer gene complex region.
Spécification
| CLEC4E | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% BSA and no preservative | |
| Q67EQ1 | |
| CLEC4E | |
| Rat mincle-Fcgamma2b fusion protein. | |
| 100 μg | |
| Primary | |
| Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Functional Assay | |
| WEN43 | |
| Unconjugated | |
| CLEC4E | |
| C86253; CLEC4E; CLECSF9; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 9; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 9; C-type lectin domain family 4 member E; C-type lectin domain family 4, member E; C-type lectin superfamily member 9; C-type lectin, superfamily member 9; immunoreceptor; macrophage-inducible C-type lectin; Mincle; UNQ218/PRO244 | |
| Mouse | |
| Protein G | |
| RUO | |
| 450223 | |
| 4°C | |
| Liquid |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.