Learn More
CLIP-115/CLIP2 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 67776-1-IG-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The CLIP-115/CLIP2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CLIP-115/CLIP2. This antibody recognizes human, mouse, rat, pig, rabbit antigen. The CLIP-115/CLIP2 antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Spécification
| CLIP-115/CLIP2 | |
| Monoclonal | |
| 1000 μg/mL | |
| CLIP; CLIP-115; CYLN2; KIAA0291; MGC11333; WBSCR3; WBSCR4; WSCR3; WSCR4 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat, Pig, Rabbit | |
| 120 kDa | |
| Liquid |
| Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
| 2E6B6 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 7461 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |