missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CoAA Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA5115829
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Antibody detects endogenous levels of total RBM14.
Isoform 1 may function as a nuclear receptor coactivator, enhancing transcription through other coactivators such as NCOA6 and CITED1. Isoform 2, functions as a transcriptional repressor, modulating transcriptional activities of coactivators including isoform 1, NCOA6 and CITED1.
Spezifikation
| CoAA | |
| Polyclonal | |
| Unconjugated | |
| RBM14 | |
| 1300007E16Rik; COAA; coactivator activator; p16; p16K; Paraspeckle protein 2; PSP2; Rbm14; RNA binding motif protein 14; rna-binding; RNA-binding motif protein 14; RNA-binding protein 14; RRM-containing coactivator activator/modulator; SIP; synaptotagmin interacting protein; synaptotagmin-interacting protein; SYT-interacting protein; Sytip; SYTIP1; TMEM137; transmembrane protein 137; Unknown (protein for MGC:137891) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10432, 170900, 56275 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q8C2Q3, Q96PK6 | |
| RBM14 | |
| A synthesized peptide derived from human RBM14(Accession Q96PK6), corresponding to amino acid residues G545-A595. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur