missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coatomer protein complex, subunit zeta 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-30435-25ul
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Coatomer protein complex, subunit zeta 2 Polyclonal specifically detects Coatomer protein complex, subunit zeta 2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Spécification
| Coatomer protein complex, subunit zeta 2 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Q9P299 | |
| COPZ2 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: DPQQVIQKVNFRADDGGLTEQSVAQV | |
| 25ul | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| coatomer protein complex, subunit zeta 2, coatomer subunit zeta-2, MGC23008, nonclathrin coat protein zeta-COP, zeta-2 coat protein, Zeta-2 COP, Zeta-2-coat protein, zeta2-COP | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 51226 | |
| Human | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu