missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Collagen V Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA5115037
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Antibody detects endogenous levels of total Collagen V alpha 1.
COL5A1 encodes an alpha chain for one of the lowabundance fibrillar collagens. Fibrillar collagen molecules aretrimers that can be composed of one or more types of alpha chains.Type V collagen is found in tissues containing type I collagen andappears to regulate the assembly of heterotypic fibers composed ofboth type I and type V collagen. COL5A1 product is closelyrelated to type XI collagen and it is possible that the collagenchains of types V and XI constitute a single collagen type withtissue-specific chain combinations. Mutations in this gene areassociated with Ehlers-Danlos syndrome, types I and II.
Spécification
| Collagen V | |
| Polyclonal | |
| Unconjugated | |
| COL5A1 | |
| AB collagen; AI413331; alpha-1 type V collagen; alpha1(V) collagen; COL5; Col5a1; Collagen 5; collagen alpha 1 (V); collagen alpha-1(V) chain; collagen type 5 alpha 1; collagen type V alpha 1; collagen type V alpha 1 chain; collagen, type V, alpha 1; EDSC; pro-alpha1(V) collagen; procollagen alpha 1(V); procollagen type V alpha 1; procollagen, type V, alpha 1; RP11-263F14.1; Type V Collagen | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1289 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P20908 | |
| COL5A1 | |
| A synthesized peptide derived from human COL5A1(Accession P20908), corresponding to amino acid residues G874-R924. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu