Learn More
Collagen VII Antibody (LH7.2), Alexa Fluor™ 350, Novus Biologicals™
Tous les produits Bio Techne ProduitsDescription
Collagen VII Monoclonal antibody specifically detects Collagen VII in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Spécification
Spécification
| Antigène | Collagen VII |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Immunogène | Cells from a single cell suspension of epidermal cells (obtained from fresh human neonatal foreskin) were lysed with Nonidet P40 in phosphate buffered saline and the insoluble pellet was sonicated to prepare insoluble fractions. |
| Contenu et stockage | Store at 4C in the dark. |
| Isotype | IgG1 κ |