missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COLQ Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-84715-0.1ML
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
COLQ Polyclonal specifically detects COLQ in Human samples. It is validated for Western Blot.
Spécification
| COLQ | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 8292 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| acetylcholinesterase collagenic tail peptide, AChE Q subunit, collagenic tail of endplate acetylcholinesterase, collagen-like tail subunit (single strand of homotrimer) of asymmetricacetylcholinesterase, EADAcetylcholinesterase-associated collagen, FLJ55041, single strand of homotrimeric collagen-like tail subunit of asymmetricacetylcholinesterase | |
| The immunogen is a synthetic peptide directed towards the middle region of human COLQ | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu