missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Complement C5 Antibody (eculizumab) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Marque: Novus Biologicals™ NBP3-28756-100ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Complement C5 Monoclonal antibody specifically detects Complement C5 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Spécification
| Complement C5 | |
| Monoclonal | |
| Unconjugated | |
| P01031 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| eculizumab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| anaphylatoxin C5a analog, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4, C5, complement C5, complement component 5, CPAMD4MGC142298, FLJ17816, FLJ17822 | |
| Complement C5 | |
| 100 μg | |
| Immunology, Innate Immunity, Stem Cell Markers | |
| 727 | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu