missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Complement C9 Monoclonal Antibody (C9-6-25-5)

Rat Monoclonal Antibody
Marque: Invitrogen™ MA549001
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Complement C9 Monoclonal Antibody for Western Blot
C5b-9 membrane attack complexes are assembled from five precursor molecules in the serum. Proteolytic cleavage of C5 by C5 convertase generates C5b which initiates assembly of the C5b-9 complex. The last step of C5b-9 complex formation involves polymerization of C9 which accompanies insertion of the complex into the cell membrane. During formation of C5b-8 and C9 polymerization, neoantigens are generated which are unique to the C5b-9 complex and are not present on any of the individual native complex components. The complement regulatory proteins CD59 and complement S-protein can both prevent C5b-9 insertion into the cell membrane. The formed SC5b-9 complex is unable to attach to cells and is cytolytically inactive. C5b-9 is involved in the progression of chronic proteinuric renal disease by mediating continuous tubulointerstitial damage. Early tubulointerstitial injury in the remnant kidney can be improved when C5b-9 complex forming is abrogated.
Spécification
| Complement C9 | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% BSA and 0.02% sodium azide | |
| P06683 | |
| C9 | |
| Murine C9 peptide. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Western Blot | |
| C9-6-25-5 | |
| Unconjugated | |
| C9 | |
| ARMD15; C9; C9D; complement C9; complement component 9; complement component C9; Complement component C9a; Complement component C9b; complement protein C9; complement protein C9 precursor | |
| Rat | |
| Protein G | |
| RUO | |
| 12279 | |
| 4°C | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu