Learn More
COPG Antibody, Novus Biologicals™
Code produit 18680006 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
100 μg
50 μg
Conditionnement:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18680006 100 μg 100µL
18601146 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 18680006 Fournisseur Novus Biologicals Code fournisseur NBP299186100ul
Rabbit Polyclonal Antibody
COPG Polyclonal antibody specifically detects COPG in Human samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Spécification
Antigène COPG
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugué Unconjugated
Dilution Western Blot 1:500-1:1000, Immunoprecipitation 0.2-1 uL/mg of lysate
Formule 0.2 um filtered solution in PBS
Alias de gène coat protein gamma-cop, coatomer protein complex, subunit gamma, coatomer protein complex, subunit gamma 1, COPG1coatomer subunit gamma, FLJ21068, Gamma-coat protein, gamma-COP
Espèces hôtes Rabbit
Immunogène Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human COPG.
Méthode de purification Antigen and protein A Affinity-purified
Quantité 100 μg
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 22820
Espèces cibles Human
Contenu et stockage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Forme Purified
Isotype IgG