Learn More
CoraLite™ Plus 488-conjugated C3/C3b/C3c Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech CL488-66157-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The C3/C3b/C3c antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human C3/C3b/C3c. This antibody recognizes human antigen. The C3/C3b/C3c antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.Spécification
| C3/C3b/C3c | |
| Monoclonal | |
| 1000 μg/mL | |
| ARMD9; ASP; CPAMD1 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| 115 kDa | |
| Liquid |
| Immunocytochemistry/Immunofluorescence | |
| 2A6B3 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 718 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG1 |