missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated SMO Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech CL488-66851-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The SMO antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SMO. This antibody recognizes human, canine antigen. The SMO antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.Spécification
| SMO | |
| Monoclonal | |
| 2000 μg/mL | |
| Gx; SMOH | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Canine | |
| Conjugated Antibody | |
| IgG2b |
| Immunocytochemistry/Immunofluorescence | |
| 2B10A11 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 6608 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto